Counting chambers, BLAUBRAND® Neubauer Improved
Supplier: Brand
717805 717820
BRND717805EA 76.2 EUR
BRND717805 BRND717820
Counting chambers, BLAUBRAND® Neubauer Improved
Slides Counting Chambers
The ruling shows nine large squares of 1 mm². The marked 'L' 4 large squares in the corners are divided into 16 squares with 0,25 mm sides. They are used for counting leucocytes.
  • The large square in the middle is subdivided into 25 squares each with a side length of 0,2 mm
  • Each group square consists of 16 smaller squares each with 0,05 mm sides and an area of 0,0025 mm²
  • The five group squares in the central large square are used for platelets and red blood cell count
Deserves special attention that all group squares have triple boundary lines. TheMore Product Information