Graduated pipettes, BLAUBRAND®, USP, type 2, Class AS, total delivery
Supplier: Brand
27509 27506 27511 27513 USP27511 USP27506 USP27513 USP27509
612-4755EA 110 EUR
612-4755 612-4754 612-4756 612-4757 BRNDUSP27511 BRNDUSP27506 BRNDUSP27513 BRNDUSP27509
Graduated pipettes, BLAUBRAND®, USP, type 2, Class AS, total delivery
Pipettes Graduated Pipettes
AR-Glas®
  • Calibrated to deliver (TD, Ex), nominal volume at the top
  • Marks and inscriptions in high contrast blue enamel
  • Waiting time 5 seconds
  • Suitable for cotton plugs
  • Incl. USP batch certificate, on request also available with USP individual certificate
DIN EN ISO 835
Length: 360 ±10 mm
Certifications: DIN EN ISO 835. DE-M marking.
Caution: * Error limit USP: ±0,01 ml.