Anti-Contactin-6 Rabbit Polyclonal Antibody
Supplier: Biosensis
R-127-100
BSENR-127-100EA 302 EUR
BSENR-127-100
Anti-Contactin-6 Rabbit Polyclonal Antibody
Antibodies
The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development. Employing Northern blot analysis Kamei et al found that amongst different regions of the adult human nervous system cerebellum expressed highest level of NB-3 mRNA. The expression of NB-3 in the cerebellum increases until adulthood. In contrast, the expression in the cerebrum declines to a low level after postnatal day 7. NB-3 likeMore Product Information