Anti-Ubiquitin Rabbit Polyclonal Antibody
Supplier: Biosensis
R-1405-50
BSENR-1405-50EA 302 EUR
BSENR-1405-50
Anti-Ubiquitin Rabbit Polyclonal Antibody
Antibodies
Ubiquitin is a highly conserved 76 amino acid protein with an estimated molecular weight of 8.56 kDa which has a central role in regulated protein degradation. It is a protein modifier which can be covalently attached to target lysines either as a monomer or as a lysine-linked polymer. Several types of polymeric chains can be formed depending on the lysine used for the assembly. Attachment to proteins as a polymer leads to their degradation by the 26S proteosome; a complex, multicatalytic cytosolic and nuclear protease. Attachment to proteins as a monomer or as an alternatively linkedMore Product Information