Anti-the Tyrosine Kinase Receptor B ECD Rabbit Polyclonal Antibody
Supplier: Biosensis
R-149-100
BSENR-149-100EA 302 EUR
BSENR-149-100
Anti-the Tyrosine Kinase Receptor B ECD Rabbit Polyclonal Antibody
Antibodies
TrkB is a member of the neurotrophic tyrosine receptor kinase family. It is a membrane-bound receptor and upon neurotrophin binding, it phosphorylates itself as well as MAPK pathways members. TrkB is the receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5 but not nerve growth factor (NGF). It is Involved in the development and/or maintenance of the nervous system. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures.  SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein.More Product Information