Anti-Peroxiredoxin-6 Rabbit Polyclonal Antibody
Supplier: Biosensis
R-169-100
BSENR-169-100EA 312 EUR
BSENR-169-100
Anti-Peroxiredoxin-6 Rabbit Polyclonal Antibody
Antibodies
FUNCTION: Involved in redox regulation of the cell. Can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. May play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury.  SUBUNIT: Homotetramer. SUBCELLULAR LOCATION: Cytoplasm. Lysosome. Also found in lung secretory organelles. MISCELLANEOUS: The active site is the redox-active Cys-47 oxidized to Cys-SOH. Cys-SOH may rapidly react with a Cys-SH of the other subunit to form an intermolecular disulfide with a concomitant homodimer formation. The enzyme may beMore Product Information