High temperature ovens, forced convection, LHT series
Supplier: CARBOLITE
LHT4120301 LHT41203216P1 LHT41203508P1 LHT430301 LHT4303216P1 LHT4303508P1 LHT460301 LHT4603216P1 LHT4603508P1 LHT5120301 LHT51203216P1 LHT51203508P1 LHT530301 LHT5303216P1 LHT5303508P1 LHT560301 LHT5603216P1 LHT5603508P1 LHT6120301 LHT61203216P1 LHT61203508P1 LHT630301 LHT6303216P1 LHT6303508P1 LHT660301 LHT6603216P1 LHT6603508P1 LHT4120-230SN LHT4030-230SN LHT4060-230SN LHT5120-230SN LHT5030-230SN LHT5060-230SN LHT6120-230SN LHT6030-230SN LHT6060-230SN
466-0218EA 9020 EUR
466-0218 CARBLHT41203216P1 CARBLHT41203508P1 466-0220 CARBLHT4303216P1 CARBLHT4303508P1 466-0219 CARBLHT4603216P1 CARBLHT4603508P1 466-0215 CARBLHT51203216P1 CARBLHT51203508P1 466-0217 CARBLHT5303216P1 CARBLHT5303508P1 466-0216 CARBLHT5603216P1 CARBLHT5603508P1 466-0212 CARBLHT61203216P1 CARBLHT61203508P1 466-0214 CARBLHT6303216P1 CARBLHT6303508P1 466-0213 CARBLHT6603216P1 CARBLHT6603508P1 CARBLHT4120-230SN CARBLHT4030-230SN CARBLHT4060-230SN CARBLHT5120-230SN CARBLHT5030-230SN CARBLHT5060-230SN CARBLHT6120-230SN CARBLHT6030-230SN CARBLHT6060-230SN
High temperature ovens, forced convection, LHT series
Ovens High Temperature Ovens
Ovens have a stainless steel inner chamber which is resistant to chemical attack and a corrosion resistant mild steel exterior, with a stoved epoxy/polyester coating that is easy to clean and scratch resistant. All units have multi position shelving for convenient loading/unloading. Low thermal mass insulation materials are used throughout in order to achieve rapid heating and to allow efficient energy utilisation. The high performance fan ensures optimum temperature uniformity throughout the work chamber.
  • Choice of models from 30 to 120 litres, operating from ambient +10 to 400, 500
More Product Information