High temperature furnaces, RHF
Supplier: CARBOLITE
RHF163301 RHF168301 RHF143301 RHF148301 RHF1415301 RHF153301 RHF158301 RHF1515301 RHF1615301 RHF1683508P1 RHF1683216P1 RHF14153216P1 RHF14153508P1 RHF1433216P1 RHF1433508P1 RHF1483216P1 RHF1483508P1 RHF15153216P1 RHF15153508P1 RHF1533216P1 RHF1533508P1 RHF1583216P1 RHF1583508P1 RHF16153216P1 RHF16153508P1 RHF1633216P1 RHF1633508P1 RHF1603-400TN RHF1608-230SN RHF1608-400TN RHF1415-230SN RHF1415-400TN RHF1403-230SN RHF1403-400TN RHF1408-230SN RHF1408-400TN RHF1515-230SN RHF1515-400TN RHF1503-230SN RHF1503-400TN RHF1508-230SN RHF1508-400TN RHF1615-230SN RHF1615-400TN RHF1603-230SN
455-0044EA 9950 EUR
455-0044 455-0045 455-0038 455-0039 455-0037 455-0041 455-0042 455-0040 455-0043 CARBRHF1683508P1 CARBRHF1683216P1 CARBRHF14153216P1 CARBRHF14153508P1 CARBRHF1433216P1 CARBRHF1433508P1 CARBRHF1483216P1 CARBRHF1483508P1 CARBRHF15153216P1 CARBRHF15153508P1 CARBRHF1533216P1 CARBRHF1533508P1 CARBRHF1583216P1 CARBRHF1583508P1 CARBRHF16153216P1 CARBRHF16153508P1 CARBRHF1633216P1 CARBRHF1633508P1 CARBRHF1603-400TN CARBRHF1608-230SN CARBRHF1608-400TN CARBRHF1415-230SN CARBRHF1415-400TN CARBRHF1403-230SN CARBRHF1403-400TN CARBRHF1408-230SN CARBRHF1408-400TN CARBRHF1515-230SN CARBRHF1515-400TN CARBRHF1503-230SN CARBRHF1503-400TN CARBRHF1508-230SN CARBRHF1508-400TN CARBRHF1615-230SN CARBRHF1615-400TN CARBRHF1603-230SN
High temperature furnaces, RHF
Ovens Furnaces (Ashing and Muffle Ovens)
The RHF range of furnaces are ideal for heat treatment applications, often associated with technical ceramics. This range of higher temperature laboratory furnaces feature silicon carbide heating elements located on either side of the chamber and hard wearing refractory brick around the door and hearth, which provides rapid heating and good resistance to abrasion. Safe operation is ensured by a double skin construction providing natural air convection for low outer case temperatures and power to the elements is isolated as soon as the door is opened.
Delivery information: Supplied asMore Product Information