DSS (Disuccinimidyl suberate)

Supplier: CAYMAN

13008-50 13008-100 13008-250 13008-500
CAYM13008-50EA 18.3 EUR
CAYM13008-50 CAYM13008-100 CAYM13008-250 CAYM13008-500
DSS (Disuccinimidyl suberate)
DSS (Disuccinimidyl suberate)
Formula: C₁₆H₂₀N₂O₈
MW: 368.34 g/mol
MDL Number: MFCD00049059
CAS Number: 68528-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR