Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-2]

Supplier: Cayman Chemical
170150-100 170150-500
CAYM170150-100EA 529 EUR
CAYM170150-100 CAYM170150-500
Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-2]
Antibodies
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: PO-2
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Atlantic Cod, Atlantic Salmon, brown Trout, Carp, Flounder, Rainbow Trout, Sea Bream, Tilapia, Turbot, Wrasse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR