Gamma radiation shields
Supplier: CLEAVER SCIENTIFIC
CSR-S4G CSR-S4T-GAMMA
CLEVCSR-S4GEA 4150 EUR
CLEVCSR-S4G CLEVCSR-S4T-GAMMA
Gamma radiation shields
Shields Radiation Shields
Acrylic, transparent shield provides clear non-distorted vision, reducing light images and shadows.
  • 15° angled hourglass shape
  • Curved sides provide the user with easier access to the central area behind the shield
  • Available with curved or flat base
Thickness: 12 mm
Order Now
SPECIFICATIONS
Environmentally Preferable