Cleanroom wipes, Amplitude™ Delta™
Supplier: CONTEC
AMDE0001 AMDE0004 AMDE0003 AMDE0002
115-1850EA 896 EUR
115-1850 115-1852 115-1851 CONTAMDE0002
Cleanroom wipes, Amplitude™ Delta™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Made of 100% synthetic fibre and formed using the spunlacing process, this wipe - designed for cleaning delicate optical products such as lenses and mirrors - is exceptionally clean and durable. It has excellent chemical resistance and is therefore ideal for use with solvents. Its open fibre structure provides excellent sorptive capacity.
  • Very low levels of particles and extractables
  • Strong and durable
  • Very soft, non abrasive texture
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.
Colour: White
Caution: Data accurate at time of publishing,More Product Information