Supelco® TraceCERT® Organic Reference Standard, Terbufos-sulfone
Supplier: SUPELCO
CRM67645-50MG
CRM67645-50MGEA 69.9 EUR
CRM67645-50MG
Supelco® TraceCERT® Organic Reference Standard, Terbufos-sulfone
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P260 - P262 - P264 - P280 - P302 + P352 + P310 - P304 + P340 + P310