Disposable particle filtering respirators, FFP1/FFP2/FFP3, 5000 series
Supplier: HONEYWELL SAFETY
1005580 1005113 1005582 1005584 1005586-V2 1005099 1005602-V2 1005588 1005593 1005591 1005595 1005598 1005600 1005122
1005580.EA 31.5 EUR
1005580. 111-3547 1005582. 1005584. 1005586. 111-3563 1005602. 1005588. 1005593. 1005591. 1005595. 1005598. 1005600. DALL1005122
Disposable particle filtering respirators, FFP1/FFP2/FFP3, 5000 series
Respirators Disposable Respirators Particulate Respirators
  • Ultra-soft seal providing a soft textile sensation on the skin, absorbing perspiration, ensuring a good seal; hypoallergenic qualities
  • 'Butterfly' exhalation valve offering maximum frontal protection on the valve membrane while also providing minimal exhalation resistance
  • Latex-free, PVC-free and silicone-free materials
  • Dolomite-tested: Test demonstrating the respirator's resistance to clogging over time; ensures better breathing comfort and less worker fatigue
Colour codes: FFP1D - yellow, FFP2D - green, FFP3D - orange, FFP1D-OV - brown for organicMore Product Information