Equipment Bag
Supplier: HONEYWELL SAFETY
1007006
DALL1007006EA 42.9 EUR
DALL1007006
Equipment Bag
Bags Equipment Bags and Instrument Cases
  • Material: PVC
  • Dimensions: 500×270 mm
  • Weight: 0,600 kg
Order Now
SPECIFICATIONS
Environmentally Preferable