Pedal Stand for MAUSER WIVA Vat-5 Medical Containers
Supplier: DANIELS HEALTHCARE LIMITED
UK80800130
DANIUK80800130EA 748 EUR
DANIUK80800130
Pedal Stand for MAUSER WIVA Vat-5 Medical Containers
Retort Stands
This mobile holder is suitable for MAUSER WIVA Infinity and WIVA Vat-5 containers of 30, 50 and 60 L.
  • Made of stainless steel
  • Comes with four rollers, including two lockable
Height with open lid: 1120 mm