Humidity indicator cards
Supplier: DESCO
3HIC125 4HIC100 6HIC200
DESC3HIC125EA 33.3 EUR
DESC3HIC125 129-2642 129-2644
Humidity indicator cards
Test Papers Indicator Papers
Humidity indicator cards (HICs) are printed with moisture sensitive spots which respond to various levels of humidity with a visible colour change from blue to pink.
  • Accuracy based on ambient temperature of 75 °F
  • Tolerance is approximately 2,5% for each 10 °F away from 75 °F
The moisture sensitive spots are made of blotting paper impregnated with cobaltous chloride. The humidity inside dry packed barrier bags can be monitored by including an HIC inside. When the bag is opened, the card can be examined for proper dryness inside the bag.