Hand disinfectant gels, Soft Care Med H5
Supplier: Diversey
100858312
DIVY100858312EA 0 EUR
DIVY100858312
Hand disinfectant gels, Soft Care Med H5
Skincare Skin Disinfectants Gel Skin Disinfectants
Soft Care MED is an alcohol gel for hand disinfection. These can be used as a disinfectant for the skin, based on a mixture of isopropyl alcohol and n-propanol. This alcohol composition is a highly effective disinfectant, and extremely effective against skin-borne microorganisms (bacteria, fungi and viruses).
  • Suitable for hygienic and surgical hand disinfection
  • Contains a mixture of isopropyl alcohol and n-propanol
  • Without colour and perfume
  • Contains moisturising substances - prevents dry and irritated skin
Soft Care MED contains thickener to increaseMore Product Information