Pipette controller, Pipet-Aid® XP
Supplier: DRUMMOND
4-000-201 4-000-202 4-000-201-E 4-000-102 4-000-101
DRUM4-000-201EA 1430 EUR
DRUM4-000-201 613-4253 DRUM4-000-201-E DRUM4-000-102 DRUM4-000-101
Pipette controller, Pipet-Aid® XP
Pipettes Pipette Controllers
This lightweight, compact controller is specifically designed to minimize operator fatigue. It accepts most pipettes from 1 to 100 ml.
  • Three-speed settings provide more precise control for both aspirate and dispense modes
  • Easy access battery (9 V (Ni-MH)) replacement
  • EZ Grip nosepiece with double membrane filter
  • Small and lightweight
Each aspiration and dispense button has three speeds, (F) Fast, (M) Medium, (S) Slow. The medium speed is suitable for most routine techniques. Fast speed can deliver up to 50 ml in 5 seconds and can be used for rapidMore Product Information