Safety underbench cabinets, type 90, UTS ergo line ST, 600 mm depth
Supplier: DUPERTHAL
29-060669-501 29-060669-401 29-060669-001 29-060667-001 29-060667-001L 29-060667-501 29-060667-401
DUEH29-060669-501EA 1730 EUR
DUEH29-060669-501 DUEH29-060669-401 135-1897 DUEH29-060667-001 DUEH29-060667-001L DUEH29-060667-501 135-1895
Safety underbench cabinets, type 90, UTS ergo line ST, 600 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information