Safety underbench cabinets, Type 90, UTS ergo line MT, 600 mm depth
Supplier: DUPERTHAL
29-060967-001 29-060967-401 29-060967-501 29-060969-001 29-060969-401 29-060969-501
135-1898EA 2240 EUR
135-1898 135-1900 DUEH29-060967-501 DUEH29-060969-001 DUEH29-060969-401 DUEH29-060969-501
Safety underbench cabinets, Type 90, UTS ergo line MT, 600 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information