Ultrasonic Baths, Elmasonic Selects
Supplier: ELMA SCHMIDBAUER
1106668 1106989 1106990 1106991 1106992 1106993 1106853
ELMA1106668EA 1150 EUR
ELMA1106668 ELMA1106989 ELMA1106990 ELMA1106991 ELMA1106992 ELMA1106993 ELMA1106853
Ultrasonic Baths, Elmasonic Selects
Ultrasonic Baths
Powerful and individual - the versatile Elmasonic Select series for ultrasonic cleaning are equipped with 37 kHz ultrasonic frequency, five ultrasonic modes, the ability to save up to four cleaning programmes, a heating and tank volumes from 3 to 90 L (model Select 60 and larger have a drain).
  • Adjustable cleaning programmes
  • Memory for four individual cleaning programmes
  • Cleaning process in the ultrasonic cleaning device can be called up for reproducible results
  • Fast switching between favourite programmes
  • Auto-degas mode removes air bubbles in the cleaning
More Product Information