pH/conductivity electrode, Eutech
Supplier: EUTECH
ECCOMBI03M
EUTEECCOMBI03MEA 286 EUR
EUTEECCOMBI03M
pH/conductivity electrode, Eutech
Electrodes Multi-Parameter Electrodes
Combined pH electrode and 2-pin stainless steel conductivity electrode with 15 cm ABS guard. For use with CyberScan PC 10.