Fast red TR salt (1,5-naphthalenedisulphonate salt), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning


M-Clarity™ quality level = MQ200

F6760-1G F6760-5G
F6760-1GEA 86 EUR
F6760-1G F6760-5G
Fast red TR salt (1,5-naphthalenedisulphonate salt), Sigma-Aldrich®
Fast Red TR salt (1,5-naphthalenedisulphonate salt)

Fast Red TR Salt 1,5-naphthalenedisulfonate salt has been used for tartrate-resistant acid phosphatase (TRAP) staining in tissue sections. It has been used as a chromogen in APAAP (alkaline phosphatase-anti-alkaline phosphatase) procedure.

Formula: C₇H₉Cl₂N
Storage Temperature: Freezer
CAS Number: 51503-28-7
EINECS: 257-247-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76217843907a5f4ac49977acbace8b16d578b624&vid=76217843907a5f4ac49977acbace8b16d578b624&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) White to Light Yellow
Appearance (Form) Powder
Proton NMR spectrum Conforms to Structure
Solubility (Turbidity) Clear to Slightly Hazy
Solubility (Colour), c = 25 mg/ml, H₂O, Solvent Content (by GC) Faint Yellow to Light Yellow-Green
Suitability, Suitable as Coupling Agent with Naphthol AS Suitable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR