PolyCHECK® Wipes
Supplier: FOAMTEC
HT5291K-5
FOAMHT5291K-5EA 4800 EUR
FOAMHT5291K-5
PolyCHECK® Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
PolyCHECK® Wipe, black polyester inspection wipe with sealed edge.