VACUETTE® Urine Collection Tubes
Supplier: GREINER BIO ONE
455007_1200 456007_1200 455028_1200
GREI455007_1200EA 220 EUR
GREI455007_1200 GREI456007_1200 GREI455028_1200
VACUETTE® Urine Collection Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
These sterile tubes are used for urine collection, transportation, centrifugation, strip tests, analysis and storage.
  • Exact dosage vacuum and meet quality standards
  • With or without preservative
Hermetically sealed, unbreakable and space saving, with improved safety for laboratory and hospital personnel through the use of a closed system, contamination is virtually impossible when decanting into a secondary vessel.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904