Competent cells, Z-Competent™ E. coli transformation kits
Supplier: G-Biosciences
GZ-4 GZ-5 GZ-5S
GZ-4EA 500 EUR
GZ-4 GZ-5 GZ-5S
Competent cells, Z-Competent™ E. coli transformation kits
Competent Cells
The Z-Competent™ E. coli transformation kit is designed to generate competent E. coli cells for simple and highly efficient E. coli transformation.
  • No heat shock required
  • Competent cells in <20 mins
  • Transformations in <5 mins
  • High efficiency transformations
E. coli are grown in SOB medium (supplied with GZ-5), washed and suspended in supplied competent buffer. The bacterial cells are now ready for transformations. The transformation efficiency is 2×10⁸ to 1×10⁹ transformants per μg of pUC19 plasmid. The efficiency of transformation variesMore Product Information