Palladium 5% on activated carbon paste (50% water) 5R437

Supplier: ALFA AESAR
Warning

H33537.03 H33537.09 H33537.18
H33537.03EA 31.4 EUR
H33537.03 H33537.09 H33537.18
Palladium 5% on activated carbon paste (50% water) 5R437
Palladium

CAS No.: 7440-05-3 UN No.: UN1325 MDL No.: MFCD03457879

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR