Methyllithium 1.6M in diethyl ether

Supplier: ALFA AESAR
Danger


Packaged under Nitrogen in resealable AcroSeal® bottles

H36516.9Z H36516.AE
H36516.9ZEA 237 EUR
H36516.9Z H36516.AE
Methyllithium 1.6M in diethyl ether
Methyllithium
Formula: CH₃Li
Flash Pt: –17°C (1°F)
Storage Temperature: Ambient
MDL Number: MFCD00008253
CAS Number: 917-54-4
EINECS: 213-026-4
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR