m-FC with rosolic acid broth
Supplier: HACH
2428520
HACH2428520EA 73.5 EUR
HACH2428520
m-FC with rosolic acid broth
Media for Microbiology Prepared Media for Microbiology
Selectivity: Faecal coliform bacteria. For drinking water, wastewater, ambient water, food, and beverage applications.
  • Save valuable time and money using this prepared media for membrane filtration methods
  • Procedures, prepared media, and required consumables available for presumptive and confirming tests