1,5-Diphenylcarbazone

Supplier: HACH

96799
HACH96799EA 48.3 EUR
HACH96799
1,5-Diphenylcarbazone
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
Boiling Pt: 440 °C (1013 hPa)
Melting Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5
EINECS: 208-698-0
Merck Index: 12,03380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR