Ear punches, IMS
Supplier: HARVARD APPARATUS
34-0136
HARV34-0136EA 30.1 EUR
HARV34-0136
Ear punches, IMS
Biopsy Punches
This easily operated hole punching device is suitable for marking small rodents whenever a permanent identification system is required. Two types of punches are available. A forceps type which provides a longer handle for gripping, or the simple thumb type.
Packaging: These punches are supplied individually.