Sieve shakers, Tyler Ro-Tap®
Supplier: HAVERBOECKER
205322655UK
510-0945EA 5730 EUR
510-0945 510-0946 510-1035 510-1034 HAVE205322655UK HAVE205322761UK
Sieve shakers, Tyler Ro-Tap®
Shakers Sieve Shakers
For dry sieving. Tyler Ro-Tap® shakers RX 29 CE and RX 30 CE reproduce the circular and tapping motion of hand sieving, but in a repeatable mechanical action. This sieve shaker makes a circular movement of the sieve stack and a mechanical arm makes a tapping movement on the top of the sieve stack, thus creating a perfect and repeatable imitation of hand shaking with additional tapping of the sieve on the side. They are operated in a totally enclosed cabinet, which guarantees the safety of the operator, and reduces the sound emissions significantly.
  • Rotation frequency: 278 min⁻¹;
More Product Information