Slide box
Supplier: GLASWARENFABRIK KARL HECHT
42705010 42705002 42705003
HECH42705010EA 10.5 EUR
HECH42705010 HECH42705002 HECH42705003
Slide box
Slide Boxes
Stackable storage boxes with tight-closing lid, removable index card and numbered bottom.