Findenser™ Air condenser for Magnetic Stirrer
Supplier: Heidolph Instruments GmbH & Co.KG
505-81820-00 505-81810-00 505-81710-00 505-81720-00 505-81800-00
HEID505-81820-00EA 497 EUR
HEID505-81820-00 HEID505-81810-00 HEID505-81710-00 HEID505-81720-00 HEID505-81800-00
Findenser™ Air condenser for Magnetic Stirrer
Condensers
The high performance Findenser™ as standard size (400 mm) is for syntheses under reflux with solvent voluments up to 1 liter.
  • Save water, space and costs
  • Patented design
  • Finned aluminium jacket
  • Excellent thermal conductivity
  • High performance air cooling
  • Chemically resistant anodised surface
  • Ground glass cones
It can be secured with standard laboratory clamps or with the Findenser™ attachment kit (sold separately).
The Findenser™ is suitable for reflux cooling.
The Findenser™ may be operated in research laboratories, other laboratories andMore Product Information