Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Supplier: Heraeus
TDE30086FV5ID TDE40086FV5ID TDE50086FV5ID TDE60086FV5ID TDE30086FV5I-U TDE40086FV5I-U TDE50086FV5I-U TDE60086FV5I-U
HERATDE30086FV5IDEA 15660 EUR
HERATDE30086FV5ID HERATDE40086FV5ID HERATDE50086FV5ID HERATDE60086FV5ID HERATDE30086FV5I-U HERATDE40086FV5I-U HERATDE50086FV5I-U HERATDE60086FV5I-U
Ultra low temperature freezers, with 5 inner doors, –86 °C, TDE series
Freezers Ultra Low Temperature Freezers
Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with 5 Inner doors, maximizing storage capacity from 30000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. Sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-
More Product Information