Cone supplies
Supplier: Agilent
5188-5359
HEWL5188-5359EA 266 EUR
HEWL5188-5359
Cone supplies
Chromatography Supplies Chromatography Spare Parts
A range of accessories and supplies for skimmer and sampling cones for use with our ICP-MS portfolio.
  • Viton O-ring for sampling cone that seals the sampling cone to the vacuum chamber
  • Screws for skimmer bases are available and used to install the sampling base to the vacuum chamber
  • They also provide round interface wrenches (in order to remove and install the skimmer cone) and sampling cone wrenches (used to remove and install the sampling cone)
High quality ICP-MS systems contain interfaces that comprise of a step-down vacuum stage, located between aMore Product Information