Ethyl acetate ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™

Supplier: HONEYWELL
Danger

27227-180KG 27227-1L-GL 27227-1L 27227-2.5L 27227-35KG 27227-4X2.5L 27227-4X5L 27227-6X1L 27227-18L 27227-7L-RC
27227-180KGEA 1720 EUR
27227-180KG 27227-1L-GL 27227-1L 27227-2.5L 27227-35KG 27227-4X2.5L 27227-4X5L 27227-6X1L HONC27227-18L HONC27227-7L-RC
Ethyl acetate ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Ethyl acetate

Beilstein Number: 506104

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: –84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=0f6c525db7a2f3725affe5045040d70e0ac99352&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.5 %
Density (D 20/20) 0.898 - 0.902
Density (D 25/25) 0.894 - 0.898
Refractive index (n 20/D) 1.371 - 1.373
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.005 %
Ethanol (GC) Max. 0.1 %
Methyl compounds (as Acetic acid methyl ester) Max. 0.01 %
Reaction against H₂SO₄ complying
Related substances (GC) Max. 0.2 %
Chromatographic purity complying
Residual solvents complying
Appearance of the solution complying
Refractive Index n20/D 1.371-1.373
Vapor Pressure 73 mmHg ( 20 °C)
Explosion Limit 2.2-11.5 %, 38 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR