Ion selective meters for nitrate, LAQUAtwin NO3-11 / NO3-11C / NO3-11S
Supplier: HORIBA
3200689163 3200689162 3200689164
HORI3200689163EA 530 EUR
HORI3200689163 HORI3200689162 HORI3200689164
Ion selective meters for nitrate, LAQUAtwin NO3-11 / NO3-11C / NO3-11S
Electrochemistry Meters Ion Meters
These pocket meters directly measure nitrate ion and nirate-nitrogen concentrations in 0,3 ml sample (or 0,05 ml sample with sampling sheet B). The unique flat sensor allows measurement of viscous liquids, solids, and even powder samples. The sensor does not need to be dipped into the sample, a few drops are sufficient.
  • Automatic standard recognition, changeable high and low calibration values
  • Automatic hold / auto stable
  • Durable waterproof ABS housing, LCD with backlight
  • Auto power off after 30 minutes, low battery indicator
  • 400 hours battery life (
More Product Information