Microplate shakers, Variomag Monoshake and Teleshake
Supplier: THERMO H + P LABORTECHNIK
50094308 50094320 50095890
442-3161EA 1050 EUR
442-3161 442-0538 HPLT50095890
Microplate shakers, Variomag Monoshake and Teleshake
Shakers Microplate Shakers
Ultra flat microplate shakers with VARIOMAG drive which are ideally suited for easy integration into robotic systems. Monoshake is a directly controlled shaker, speed is adjusted by manually turning a button. Teleshake is the same as Monoshake, however it includes a RS232 interface, so the unit can be remotely controlled via control unit and software. Up to 14 units can be controlled directly and independently with only one RS232 interface. When unit is turned off, the shaker plate self centers in zero position.
  • Choice of direct or remote control via control unit and software
  • Direction
More Product Information