Cosmic calf serum, US origin, HyClone™
Supplier: GE Healthcare - Hyclone
SH30087.03 SH30087.02 SH30087.04 SH30087.04HI SH30087.03HI SH30087.02HI SH30087.03IR25 SH30087.04IR30 SH30087.04IR25
HYCLSH30087.03EA 143 EUR
HYCLSH30087.03 HYCLSH30087.02 HYCLSH30087.04 HYCLSH30087.04HI HYCLSH30087.03HI HYCLSH30087.02HI SH30087.03IR25 SH30087.04IR30 SH30087.04IR25
Cosmic calf serum, US origin, HyClone™
Sera (Serum)
HyClone™ Cosmic calf serum is fortified with iron and naturally derived components for increased performance with a broad spectrum of cell type. Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
All components are sourced in the U.S. and are traceable. Studies have shown Cosmic Calf outperforms iron-supplemented bovine calf serum and FBS in several applications, and performs exceptionally wellMore Product Information