2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS
Warning

0210092705 0210092791 0210092780 0210092783
ICNA0210092705EA 34.6 EUR
ICNA0210092705 ICNA0210092791 ICNA0210092780 ICNA0210092783
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid ≥99%, white crystalline powder
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

BES is a zwitterionic buffer that is used in biochemistry and molecular biology research in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies.


  • Presentation: White powder


A protocol has been reported for the use of BES buffered saline in the calcium phosphate mediated transfection of eukaryotic cells with plasmid DNA. BES, a sulfonic acid-containing cross-linking agent, induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes.


The effect of BES and other zwitterionic buffers on the Bicinchoninic acid (BCA) measurement of microgram quantities of protein has been studied. BES was found to diminish color development of the protein BCA complex by 30-35% at 50 mM, and by <10% at 5 mM, measuring in the 1-10 mg range of BSA against protein-BCA complexes formed in water. This color loss may result from binding of Cu2+ to BES.


BES may be employed as binding buffer in modified Eagle′s medium during the binding assay of human melanoma cells. It may be used as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.


Soluble 0.1M in water (clear, colorless solution)

Formula: C₆H₁₅NO₅S
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH ( 1% aq soln) 3 - 5
Assay (titration, dry basis) ≥99.0%
Solubility (0.1M in H₂O) Clear, Complete
Moisture ≤1.0%
pKa 6.9 - 7.3
UV A260 nm ≤0.10
UV A280 nm ≤0.08


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR