Choline chloride ≥98.0%, white crystalline powder

Supplier: MP BIOMEDICALS

0210138605 02101386.1 02101386.5 0210138601
ICNA0210138605EA 320 EUR
ICNA0210138605 ICNA02101386.1 ICNA02101386.5 ICNA0210138601
Choline chloride ≥98.0%, white crystalline powder
Choline chloride

Choline chloride is used as a substrate for choline acetyl transferase.


Choline chloride is an organic compound and a quaternary ammonium salt . It is a weak acid. Choline chloride is the salt of the naturally occurring choline, the pre-stage of the neurotransmitter acetylcholine, which is important for mnemonic and thought-processes. Choline occurs naturally in fungi, hop and kingcups and as integral part of lecithin. Choline chloride is a common food additive in animal husbandry.


Biochem/physiol Actions: The enzymatic activities of butyrylcholinesterase (BChE) and paraoxonase 1 (PON1), two serum enzymes synthesized by the liver and related with inflammation, were decreased in a sepsis animal model injected with LPS. Choline chloride administered intravenously at 20 mg/kg body weight prevents the LPS-mediated decreases in the activities of these two enzymes.

Formula: HOH₂CCH₂N(CH₃)₃Cl
Melting Pt: 302…305 °C (dec., lit.)
Density: ~ 1,205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=77b015e76f45084bc46da3249578a85652cd6802&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes Passes
Assay ≥98,0% (anhydrous basis)
Heavy Metals ≤10 ppm (as Pb)
pH 4 - 8
Residue on Ignition ≤0,1%
Water Content ≤0,5%
Solubility (10% aq soln) Clear and Colorless
1,4-Dioxane ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR