3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ~98%, yellow powder

Supplier: MP BIOMEDICALS
Warning

0210222701 02102227.1 02102227.5 0210222705 0210222710
ICNA0210222701EA 90.7 EUR
ICNA0210222701 ICNA02102227.1 ICNA02102227.5 ICNA0210222705 ICNA0210222710
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ~98%, yellow powder
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

3-(4,5-Dimethylthiazolyl-2)-2,5-Diphenyl Tetrazolium Bromide(MTT) is an histochemical/cytochemical reagent. It is converted to water-insoluble MTT-formazan of dark blue color by mitochondrial dehydrogenases of living cells, thus used for measurement of cell division.


Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.


Soluble in methyl alcohol (heat), methanol (10 mg/ml with heat - clear, yellow solution) and water (5 mg/ml - clear to very slightly hazy, yellow to bright yellow solution).


Caution: Research Use Only (RUO). Store at +4 °C, protect from light

Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Melting Pt: 189…199 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Melting Point 189 - 199 °C
UV/Vis: λmax (methanol) 376 ± 2 nm; 242 ± 2 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR