Uridine ≥99%, white powder

Supplier: MP BIOMEDICALS

0210321605 0210321610 0210321625 0210321680
ICNA0210321605EA 37.3 EUR
ICNA0210321605 ICNA0210321610 ICNA0210321625 ICNA0210321680
Uridine ≥99%, white powder
Uridine

Uridine is one of the four basic components of ribonucleic acid (RNA); the other three are adenosine, guanosine, and cytidine. Upon digestion of foods containing RNA, uridine is released from RNA and is absorbed intact in the gut. Uridine plays a role in the glycolysis pathway of galactose.
Uridine is used in treatment of bipolar disorder associated with mood swings.Uridine is used in metabolic labeling and analysis of rRNA processing.


  • Presentation: White Powder
  • Soluble in 0.1 N HCl (50 mg/mL - clear, colorless solution).
  • Storage temperature: Room Temperature

Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 160…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS Number: 58-96-8

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 160-171°C
UV/Vis: l max (water) 260-263nm
OD250/OD260 0.72-0.77
OD280/OD260 0.32-0.38


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR