L(+)-Lysine monohydrate, powder

Supplier: MP BIOMEDICALS

0219022405 0219022425 0219022480
ICNA0219022405EA 50.7 EUR
ICNA0219022405 ICNA0219022425 ICNA0219022480
L(+)-Lysine monohydrate, powder
L(+)-Lysine monohydrate

L-Lysine is an essential proteinogenic α amino acid. It contains an N-butyl amino group in the side chain, and this moiety is protonated at physiological pH. In addition, L-lysine is one of the two amino acids that are degraded to give ketone bodies (a ketogenic amino acid).


L-Lysine is used in a wide range of applications including as a supplement in cell culture media; a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14); a component of poly-lysine polymers; and a substrate for oxidation and glycation mechanism studies. L-lysine has been used with cultured human osteoblasts to study cell proliferation.


Lysine is degraded in vivo to give acetoacetyl CoA via an initial transamination with alpha-ketoglutarate. L-Lysine is an essential proteinogenic α amino acid used in a wide range of applications including as a supplement in cell culture media; a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14); a component of poly-lysine polymers; and a substrate for oxidation and glycation mechanism studies.


Soluble in 0,1 N hydrochloride (50 mg/ ml - clear, colorless solution) or water (100 mg/ml - clear, colorless solution).

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MDL Number: MFCD00151035
CAS Number: 39665-12-8

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 9.9 - 10.3
Loss on Drying ≤11.8%
Optical Rotation (c=8, 6 N HCl) +23.5° to +26.5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR