Calcium hydrogen orthophosphate, white powder

Supplier: MP BIOMEDICALS

0219380480 0219380490 0219380491
ICNA0219380480EA 28.2 EUR
ICNA0219380480 ICNA0219380490 ICNA0219380491
Calcium hydrogen orthophosphate, white powder
Calcium hydrogen orthophosphate

Calcium phosphate is used in animal feeds, as a mineral supplement, as an antacid, in the manufacturing of glass, in dental products and in fertilizers. It can be used as a dietary supplement, and as an antacid.


Caution: Research Use Only (RUO).

Formula: CaHPO₄
Melting Pt: ∼370 °C (dec., lit.)
Density: ∼2,3 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR