n-Hexadecane ≥99%, liquid

Supplier: MP BIOMEDICALS

0219521880 0219521890 0219521891
ICNA0219521880EA 66.4 EUR
ICNA0219521880 ICNA0219521890 ICNA0219521891
n-Hexadecane ≥99%, liquid
n-Hexadecane

Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.


Soluble in ether; insoluble in water.


Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)

Formula: H₃C(CH₂)₁₄CH₃
MDL Number: MFCD00008998
CAS Number: 544-76-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=90d3084bdd908b305ecf9b3c30e03cd98a407d94&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR