dNTP mix, MP Biomedicals
Supplier: MP BIOMEDICALS
11NTPMX055 11NTPMX255 11NTPMX250 11NTPMX100 11NTPMX050 11NTPMX105
ICNA11NTPMX055EA 276 EUR
ICNA11NTPMX055 ICNA11NTPMX255 ICNA11NTPMX250 ICNA11NTPMX100 ICNA11NTPMX050 ICNA11NTPMX105
dNTP mix, MP Biomedicals
Nucleic Acid Reagents Nucleotides
This dNTP mix with dATP, dCTP, dGTP, dTTP is suitable for use in PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, DNA sequencing and labeling.