Centring rings with fine filter, type KF
Supplier: Welch by Gardner Denver
701183 701191 701181 701193
ILMV701183EA 23.1 EUR
ILMV701183 181-0622 ILMV701181 ILMV701193
Centring rings with fine filter, type KF
Seals Sealing Rings
Centring rings made of stainless steel, with fine filter and perbunan O-ring.
Mesh width: 1,4 mm
Wire diameter: 0,2 mm